Assay Details
Target Gene Details
Entrez Gene ID: | 716 |
Gene Name: | complement component 1, s subcomponent |
Gene Aliases: |
- |
Location: |
Chr.12:7060676-7071032 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1S | NM_001734.3 | NP_001725.1 | ||
| NM_201442.2 | NP_958850.1 | |||
| XM_005253760.1 | XP_005253817.1 | |||
| AK290295.1 | ||||
| AM886411.1 | ||||
| BC056903.1 | AAH56903.1 | |||
| CD358820.1 | ||||
| DB257339.1 | ||||
| DB265957.1 | ||||
| J04080.1 | AAA51852.1 | |||
| M18767.1 | AAA51853.1 |
More Information
Set Membership: |
|

Genomic Map