Assay Details
Target Gene Details
Entrez Gene ID: | 10112 |
Gene Name: | kinesin family member 20A |
Gene Aliases: |
MKLP2, RAB6KIFL |
Location: |
Chr.5:138178728-138187715 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF20A | NM_005733.2 | 6 | 1099 | NP_005724.1 |
| AF070672.1 | 5 | 630 | AAC83230.1 | |
| AF153329.1 | 6 | 637 | AAD37806.1 | |
| AK025790.1 | 6 | 658 | ||
| AK296879.1 | 7 | 613 | ||
| BC012999.2 | 6 | 637 | AAH12999.1 |
More Information
Set Membership: |
|

Genomic Map