Assay Details
Target Gene Details
Entrez Gene ID: | 50604 |
Gene Name: | interleukin 20 |
Gene Aliases: |
IL-20, IL10D, ZCYTO10 |
Location: |
Chr.1:206865247-206869223 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL20 | NM_018724.3 | 5 | 1111 | NP_061194.2 |
| XM_005273152.3 | 6 | 1537 | XP_005273209.1 | |
| AY358320.1 | 6 | 1430 | AAQ88686.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2660993 | Chr.1:206867257 - 206871886 on Build GRCh38 | Deletion |
|
| esv3588662 | Chr.1:206867210 - 206871906 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map