Assay Details
Target Gene Details
Entrez Gene ID: | 130 |
Gene Name: | alcohol dehydrogenase 6 (class V) |
Gene Aliases: |
ADH-5 |
Location: |
Chr.4:99202638-99219246 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADH6 | NM_000672.3 | NP_000663.1 | ||
| NM_001102470.1 | NP_001095940.1 | |||
| NR_132990.1 | ||||
| AK092768.1 | ||||
| AK316326.1 | ||||
| BC039065.1 | AAH39065.1 |
Target Gene Details
Entrez Gene ID: | 100507053 |
Gene Name: | uncharacterized LOC100507053 |
Gene Aliases: |
- |
Location: |
Chr.4:99088857-99301356 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507053 | NR_037884.1 | |||
| BX647987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5345n100 | Chr.4:99181544 - 99523288 on Build GRCh38 | Gain |
|
| nsv1002927 | Chr.4:99031321 - 99227533 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map