Assay Details
Target Gene Details
Entrez Gene ID: | 3586 |
Gene Name: | interleukin 10 |
Gene Aliases: |
CSIF, GVHDS, IL-10, IL10A, TGIF |
Location: |
Chr.1:206767603-206772494 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL10 | NM_000572.2 | 5 | 990 | NP_000563.1 |
| M57627.1 | 5 | 961 | AAA63207.1 |
More Information
Set Membership: |
|

Genomic Map