Assay Details
Target Gene Details
Entrez Gene ID: | 269 |
Gene Name: | anti-Mullerian hormone receptor type 2 |
Gene Aliases: |
AMHR, MISR2, MISRII, MRII |
Location: |
Chr.12:53423855-53431672 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMHR2 | NM_001164690.1 | NP_001158162.1 | ||
| NM_001164691.1 | NP_001158163.1 | |||
| NM_020547.2 | NP_065434.1 | |||
| XM_011538173.1 | XP_011536475.1 | |||
| XM_011538174.1 | XP_011536476.1 | |||
| XM_011538176.1 | XP_011536478.1 | |||
| XM_011538178.1 | XP_011536480.1 | |||
| XM_011538179.1 | XP_011536481.1 | |||
| XM_011538180.1 | XP_011536482.1 | |||
| XM_011538181.1 | XP_011536483.1 | |||
| XM_011538183.2 | XP_011536485.1 | |||
| XM_011538184.2 | XP_011536486.1 | |||
| XM_011538185.1 | XP_011536487.1 | |||
| XM_011538186.2 | XP_011536488.1 | |||
| XM_017019179.1 | XP_016874668.1 | |||
| AF172932.1 | AAD48497.1 | |||
| AK313593.1 | ||||
| AY714878.1 | AAU21221.1 | |||
| BC126316.1 | ||||
| BC136356.1 | ||||
| CR627318.1 | 4 | 741 |
More Information
Set Membership: |
|

Genomic Map