Assay Details
Target Gene Details
Entrez Gene ID: | 55779 |
Gene Name: | cilia and flagella associated protein 44 |
Gene Aliases: |
WDR52 |
Location: |
Chr.3:113286930-113445190 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Target Gene Details
Entrez Gene ID: | 100874029 |
Gene Name: | CFAP44 antisense RNA 1 |
Gene Aliases: |
WDR52-AS1 |
Location: |
Chr.3:113403998-113433992 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP44-AS1 | NR_046728.1 | |||
| BX108680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013506 | Chr.3:113171761 - 113962104 on Build GRCh38 | Gain |
|

Genomic Map