Assay Details
Target Gene Details
Entrez Gene ID: | 54838 |
Gene Name: | WW domain binding protein 1-like |
Gene Aliases: |
C10orf26, OPA1L, OPAL1 |
Location: |
Chr.10:102742708-102816267 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WBP1L | NM_001083913.1 | NP_001077382.1 | ||
| NM_017787.4 | NP_060257.4 | |||
| XM_017016360.1 | XP_016871849.1 | |||
| AK056285.1 | ||||
| AK300871.1 | ||||
| AK308843.1 | ||||
| AK316452.1 | ||||
| AY656171.1 | ||||
| BC113857.1 | AAI13858.1 | |||
| BC114465.1 | AAI14466.1 | |||
| CX163170.1 | ||||
| DA797839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2674595 | Chr.10:102775676 - 102781207 on Build GRCh38 | Deletion |
|
| nsv831969 | Chr.10:102651251 - 102812041 on Build GRCh38 | Loss |
|
| esv3624401 | Chr.10:102775660 - 102781271 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map