Assay Details
Target Gene Details
Entrez Gene ID: | 606495 |
Gene Name: | cytochrome b5 reductase like |
Gene Aliases: |
- |
Location: |
Chr.1:54172336-54200073 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB5RL | NM_001031672.2 | NP_001026842.2 | ||
| AK090456.1 | BAC03437.1 | |||
| CD515193.1 | ||||
| DA793739.1 |
More Information
Set Membership: |
|

Genomic Map