Assay Details
Target Gene Details
Entrez Gene ID: | 22839 |
Gene Name: | DLG associated protein 4 |
Gene Aliases: |
DAP-4, DAP4, DLP4, SAPAP-4, SAPAP4 |
Location: |
Chr.20:36306323-36528637 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLGAP4 | NM_001042486.3 | NP_001035951.1 | ||
| NM_014902.5 | NP_055717.2 | |||
| NM_183006.3 | NP_892118.1 | |||
| XM_005260332.4 | XP_005260389.1 | |||
| XM_005260333.4 | XP_005260390.1 | |||
| XM_011528688.2 | XP_011526990.1 | |||
| XM_011528689.2 | XP_011526991.1 | |||
| XM_011528691.2 | XP_011526993.1 | |||
| XM_011528692.2 | XP_011526994.1 | |||
| XM_017027725.1 | XP_016883214.1 | |||
| XM_017027726.1 | XP_016883215.1 | |||
| XM_017027727.1 | XP_016883216.1 | |||
| AB023181.1 | ||||
| AK001681.1 | ||||
| AK055152.1 | ||||
| AK125383.1 | ||||
| BC108706.1 | AAI08707.1 | |||
| BC153874.1 |
Target Gene Details
Entrez Gene ID: | 101926987 |
Gene Name: | DLGAP4 antisense RNA 1 |
Gene Aliases: |
CCAT7 |
Location: |
Chr.20:36507702-36573275 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLGAP4-AS1 | NR_109939.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833967 | Chr.20:36477427 - 36658869 on Build GRCh38 | Loss |
|
| nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map