Assay Details
Target Gene Details
Entrez Gene ID: | 4356 |
Gene Name: | membrane palmitoylated protein 3 |
Gene Aliases: |
DLG3 |
Location: |
Chr.17:43800799-43833192 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPP3 | NM_001932.4 | 20 | 2554 | NP_001923.2 |
| NR_003562.2 | 19 | 2533 | ||
| U37707.1 | 18 | 2715 | AAB36964.1 |
More Information
Set Membership: |
|

Genomic Map