Assay Details
Target Gene Details
Entrez Gene ID: | 144108 |
Gene Name: | SPT2 chromatin protein domain containing 1 |
Gene Aliases: |
Spt2 |
Location: |
Chr.11:18606401-18634823 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPTY2D1 | NM_194285.2 | 3 | 1211 | NP_919261.2 |
| XM_011519919.2 | 3 | 1069 | XP_011518221.1 | |
| AF452716.1 | 1 | 605 | AAP13351.1 | |
| AK096760.1 | 3 | 1231 | BAC04858.1 | |
| AK304078.1 | 3 | 1127 | ||
| BC056261.1 | AAH56261.1 | |||
| BX641102.1 | 3 | 1112 | CAE46047.1 | |
| BX647798.1 | 3 | 1211 | ||
| BX648114.1 | 1 | 790 | CAH10772.1 | |
| BX648478.1 | 3 | 1133 | ||
| CR749626.1 | 1 | 654 | CAH18420.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553633 | Chr.11:18290154 - 19036904 on Build GRCh38 | Gain |
|
| nsv516413 | Chr.11:18606871 - 18622211 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map