Assay Details
Target Gene Details
Entrez Gene ID: | 317781 |
Gene Name: | DEAD-box helicase 51 |
Gene Aliases: |
- |
Location: |
Chr.12:132136594-132144335 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX51 | NM_175066.3 | NP_778236.2 | ||
| XM_011538256.1 | XP_011536558.1 | |||
| AB593157.1 | ||||
| AK097078.1 | BAC04942.1 | |||
| AL079273.1 | ||||
| BC040185.2 | AAH40185.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952511 | Chr.12:132132756 - 132182155 on Build GRCh38 | Deletion |
|
| nsv1041402 | Chr.12:132076412 - 132441297 on Build GRCh38 | Gain |
|
| nsv826563 | Chr.12:132125383 - 132622340 on Build GRCh38 | Gain |
|
| nsv944 | Chr.12:132138516 - 132164483 on Build GRCh38 | Insertion |
|
| nsv509493 | Chr.12:132092623 - 132165751 on Build GRCh38 | Insertion |
|
| nsv470333 | Chr.12:132117051 - 132219256 on Build GRCh38 | Loss |
|
| nsv560935 | Chr.12:132134511 - 132150976 on Build GRCh38 | Loss |
|
| nsv428285 | Chr.12:132065104 - 132208513 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs201907915] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map