Assay Details
Target Gene Details
Entrez Gene ID: | 6943 |
Gene Name: | transcription factor 21 |
Gene Aliases: |
POD1, bHLHa23 |
Location: |
Chr.6:133889121-133895537 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCF21 | NM_003206.3 | 2 | 989 | NP_003197.2 |
| NM_198392.2 | NP_938206.1 | |||
| AF035718.1 | 2 | 973 | AAC62514.1 | |
| AF047419.1 | 2 | 972 | AAC62532.1 | |
| AL550251.3 | ||||
| BC025697.1 | AAH25697.1 | |||
| BU688835.1 | 1 | 297 | ||
| W73859.1 | 2 | 361 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34122 | Chr.6:133767881 - 134057114 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map