Assay Details
Target Gene Details
Entrez Gene ID: | 3577 |
Gene Name: | C-X-C motif chemokine receptor 1 |
Gene Aliases: |
C-C, C-C-CKR-1, CD128, CD181, CDw128a, CKR-1, CMKAR1, IL8R1, IL8RA, IL8RBA |
Location: |
Chr.2:218162845-218166993 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCR1 | NM_000634.2 | 2 | 1696 | NP_000625.1 |
| AK309632.1 | ||||
| BC028221.1 | 2 | 1655 | AAH28221.1 | |
| BC072397.1 | 2 | 1645 | AAH72397.1 | |
| L19591.1 | 2 | 1696 | AAB59436.1 | |
| M68932.1 | 2 | 1627 | AAA59159.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3164 | Chr.2:218156204 - 218187521 on Build GRCh38 | Insertion |
|
| esv3594330 | Chr.2:218142447 - 218226633 on Build GRCh38 | Gain |
|
| esv3594329 | Chr.2:218142358 - 218196924 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map