Assay Details
Target Gene Details
Entrez Gene ID: | 10238 |
Gene Name: | DDB1 and CUL4 associated factor 7 |
Gene Aliases: |
AN11, HAN11, SWAN-1, WDR68 |
Location: |
Chr.17:63550435-63594279 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF7 | NM_005828.4 | 1 | 286 | NP_005819.3 |
| NR_073585.1 | 1 | 286 | ||
| AK294925.1 | 1 | 260 | ||
| AK303212.1 | 1 | 221 | ||
| BQ221019.1 | 1 | 246 | ||
| DA230140.1 | 1 | 260 | ||
| HY023965.1 | 1 | 287 | ||
| U94747.1 | 1 | 115 | AAC18913.1 |
More Information
Set Membership: |
|

Genomic Map