Assay Details
Target Gene Details
Entrez Gene ID: | 92799 |
Gene Name: | SH3KBP1 binding protein 1 |
Gene Aliases: |
PP203, Sb1 |
Location: |
Chr.19:40576851-40591399 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHKBP1 | NM_138392.3 | NP_612401.2 | ||
| XM_006723474.1 | XP_006723537.1 | |||
| XM_006723475.1 | XP_006723538.1 | |||
| XM_011527492.1 | XP_011525794.1 | |||
| XM_011527493.1 | XP_011525795.1 | |||
| XM_017027473.1 | XP_016882962.1 | |||
| XM_017027474.1 | XP_016882963.1 | |||
| AF258553.1 | AAG23756.1 | |||
| AK075057.1 | BAC11374.1 | |||
| AK123695.1 | ||||
| AK297006.1 | ||||
| AK300808.1 | ||||
| AY007117.1 | ||||
| BC007653.1 | AAH07653.1 | |||
| BC022855.1 | AAH22855.1 | |||
| CA416700.1 |
More Information
Set Membership: |
|

Genomic Map