Assay Details
Target Gene Details
Entrez Gene ID: | 390 |
Gene Name: | Rho family GTPase 3 |
Gene Aliases: |
ARHE, Rho8, RhoE, memB |
Location: |
Chr.2:150468193-150487695 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RND3 | NM_001254738.1 | 5 | 2423 | NP_001241667.1 |
| NM_005168.4 | 6 | 2330 | NP_005159.1 | |
| BC012513.1 | 6 | 2285 | AAH12513.1 | |
| CF127383.1 | 1 | 121 | ||
| X97758.1 | 5 | 2367 | CAA66352.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834414 | Chr.2:150352860 - 150556795 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map