Assay Details
Target Gene Details
Entrez Gene ID: | 3280 |
Gene Name: | hes family bHLH transcription factor 1 |
Gene Aliases: |
HES-1, HHL, HRY, bHLHb39 |
Location: |
Chr.3:194136142-194138612 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HES1 | NM_005524.3 | NP_005515.1 | ||
| AK000415.1 | BAA91149.1 | |||
| AK300482.1 | ||||
| BC005178.1 | AAH05178.1 | |||
| BC039152.1 | AAH39152.1 | |||
| DW424342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822399 | Chr.3:194135505 - 194146719 on Build GRCh38 | Gain |
|
| nsv822397 | Chr.3:194127934 - 194147330 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map