Assay Details
Target Gene Details
Entrez Gene ID: | 5562 |
Gene Name: | protein kinase AMP-activated catalytic subunit alpha 1 |
Gene Aliases: |
AMPK, AMPKa1 |
Location: |
Chr.5:40759379-40798195 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAA1 | NM_006251.5 | 9 | 4713 | NP_006242.5 |
| NM_206907.3 | 10 | 4758 | NP_996790.3 | |
| XM_006714482.1 | 8 | 4607 | XP_006714545.1 | |
| BC037303.2 | 9 | 4695 | AAH37303.1 |
More Information
Set Membership: |
|

Genomic Map