Assay Details
Target Gene Details
Entrez Gene ID: | 55252 |
Gene Name: | additional sex combs like 2, transcriptional regulator |
Gene Aliases: |
ASXH2 |
Location: |
Chr.2:25733758-25878485 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASXL2 | NM_018263.4 | 13 | 5044 | NP_060733.4 |
| XM_006712039.3 | 9 | 7165 | XP_006712102.1 | |
| XM_006712040.2 | 9 | 4451 | XP_006712103.1 | |
| XM_011532950.2 | 12 | 5083 | XP_011531252.1 | |
| XM_011532951.2 | 11 | 4771 | XP_011531253.1 | |
| XM_017004429.1 | 13 | 4874 | XP_016859918.1 | |
| XM_017004430.1 | 10 | 7231 | XP_016859919.1 | |
| AB051472.2 | 13 | 5044 | ||
| AB084279.1 | 13 | 4907 | BAD00086.1 | |
| AK001760.1 | 1 | 2157 | BAA91889.1 | |
| BX647527.1 | 14 | 5117 | ||
| BX648218.1 | 13 | 5069 | ||
| CR933613.1 | 13 | 5088 | CAI45930.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33316 | Chr.2:25440017 - 26672800 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map