Assay Details
Target Gene Details
Entrez Gene ID: | 404216 |
Gene Name: | long intergenic non-protein coding RNA 1561 |
Gene Aliases: |
C10orf85 |
Location: |
Chr.10:120597949-120600123 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01561 | NR_103717.1 | 1 | 54 | |
| BC108662.1 | 1 | 54 | AAI08663.1 |
Target Gene Details
Entrez Gene ID: | 196051 |
Gene Name: | phospholipid phosphatase 4 |
Gene Aliases: |
DPPL2, PPAPDC1, PPAPDC1A |
Location: |
Chr.10:120456954-120600123 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPP4 | XM_005269592.2 | 6 | 9497 | XP_005269649.1 |
| XM_006717685.3 | 7 | 9337 | XP_006717748.1 | |
| XM_011539444.2 | 8 | 9395 | XP_011537746.1 | |
| XM_017015821.1 | 4 | 9078 | XP_016871310.1 | |
| XM_017015822.1 | 4 | 9045 | XP_016871311.1 | |
| DR000933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624731 | Chr.10:120590572 - 120713656 on Build GRCh38 | Gain |
|
| nsv528787 | Chr.10:120586553 - 120722450 on Build GRCh38 | Gain |
|
| esv2760128 | Chr.10:120590025 - 120720150 on Build GRCh38 | Gain |
|
| nsv1052357 | Chr.10:120592928 - 120710371 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map