Assay Details
Target Gene Details
Entrez Gene ID: | 64127 |
Gene Name: | nucleotide binding oligomerization domain containing 2 |
Gene Aliases: |
ACUG, BLAU, CARD15, CD, CLR16.3, IBD1, NLRC2, NOD2B, PSORAS1 |
Location: |
Chr.16:50693581-50733077 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOD2 | NM_001293557.1 | 3 | 1773 | NP_001280486.1 |
| NM_022162.2 | 4 | 1813 | NP_071445.1 | |
| XM_005256084.3 | 4 | 1717 | XP_005256141.1 | |
| XM_006721242.3 | 4 | 1717 | XP_006721305.1 | |
| XM_006721243.3 | 4 | 1717 | XP_006721306.1 | |
| XM_011523259.2 | 2 | 1529 | XP_011521561.1 | |
| XM_011523260.2 | 4 | 1717 | XP_011521562.1 | |
| XM_011523261.1 | 4 | 1717 | XP_011521563.1 | |
| XM_017023535.1 | 2 | 3087 | XP_016879024.1 | |
| XM_017023536.1 | 3 | 1244 | XP_016879025.1 | |
| XM_017023537.1 | 2 | 1138 | XP_016879026.1 | |
| XM_017023538.1 | 3 | 1239 | XP_016879027.1 | |
| AF178930.1 | 4 | 1813 | AAG33677.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638608 | Chr.16:50654254 - 50854693 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map