Assay Details
Target Gene Details
Entrez Gene ID: | 2247 |
Gene Name: | fibroblast growth factor 2 |
Gene Aliases: |
BFGF, FGF-2, FGFB, HBGF-2 |
Location: |
Chr.4:122826708-122898235 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF2 | NM_002006.4 | 3 | 4038 | NP_001997.5 |
| BU501243.1 | 1 | 238 | ||
| J04513.1 | 3 | 4037 | AAA52532.1 |
Target Gene Details
Entrez Gene ID: | 11162 |
Gene Name: | nudix hydrolase 6 |
Gene Aliases: |
ASFGF2, FGF-AS, FGF2AS, GFG-1, GFG1 |
Location: |
Chr.4:122892644-122923004 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT6 | NM_007083.4 | NP_009014.2 | ||
| NM_198041.2 | NP_932158.1 | |||
| AF019632.2 | AAD01635.1 | |||
| AF019633.2 | AAD01636.2 | |||
| AK291871.1 | ||||
| AK294450.1 | ||||
| BC009842.2 | AAH09842.1 | |||
| BG386740.1 | ||||
| CB128685.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894022 | Chr.4:122865924 - 123123191 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map