Assay Details
Target Gene Details
Entrez Gene ID: | 23019 |
Gene Name: | CCR4-NOT transcription complex subunit 1 |
Gene Aliases: |
AD-005, CDC39, NOT1, NOT1H |
Location: |
Chr.16:58519946-58629886 on Build GRCh38 |
Assay Gene Location: | Within Exon 50 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT1 | NM_001265612.1 | 49 | 7495 | NP_001252541.1 |
| NM_016284.4 | 49 | 7510 | NP_057368.3 | |
| NR_049763.1 | 50 | 7678 | ||
| AB023224.2 | 36 | 5555 | BAA76851.2 | |
| AF110778.1 | 9 | 1286 | AAF14861.1 | |
| AK056927.1 | 26 | 3840 | ||
| AK092885.1 | 9 | 2054 | ||
| AK301092.1 | 9 | 1274 | ||
| AK302495.1 | 26 | 3880 | ||
| AL117492.1 | 21 | 3349 | CAB55960.1 | |
| AL832769.1 | 49 | 7459 | ||
| AL833549.1 | 49 | 7469 | ||
| BC000779.2 | 11 | 1668 | AAH00779.2 | |
| BC024317.1 | 32 | 4909 | AAH24317.1 | |
| BX537840.1 | 50 | 7624 | CAD97851.1 | |
| CB162525.1 | 2 | 158 | ||
| CR749237.1 | 42 | 6448 | CAH18093.1 | |
| EF553522.1 | 49 | 7444 |
Target Gene Details
Entrez Gene ID: | 79918 |
Gene Name: | SET domain containing 6 |
Gene Aliases: |
- |
Location: |
Chr.16:58515479-58521961 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETD6 |
More Information
Set Membership: |
|

Genomic Map