Assay Details
Target Gene Details
Entrez Gene ID: | 57106 |
Gene Name: | N-acetyltransferase 14 (putative) |
Gene Aliases: |
KLP1 |
Location: |
Chr.19:55485190-55487568 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAT14 | NM_020378.3 | NP_065111.1 | ||
| AB038651.1 | BAB03716.1 | |||
| AF086533.1 | ||||
| BC019079.2 | AAH19079.1 | |||
| BM785191.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2552 | Chr.19:55467079 - 55498718 on Build GRCh38 | Insertion |
|
| nsv458833 | Chr.19:55466542 - 55521721 on Build GRCh38 | Loss |
|
| nsv953624 | Chr.19:55302033 - 55625034 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map