Assay Details
Target Gene Details
Entrez Gene ID: | 55869 |
Gene Name: | histone deacetylase 8 |
Gene Aliases: |
CDA07, CDLS5, HD8, HDACL1, MRXS6, RPD3, WTS |
Location: |
Chr.X:72329516-72573103 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC8 | NM_001166418.1 | 9 | 1604 | NP_001159890.1 |
| NM_018486.2 | 11 | 1877 | NP_060956.1 | |
| NR_051952.1 | 10 | 1735 | ||
| XM_011530986.2 | 12 | 1950 | XP_011529288.2 | |
| XM_017029640.1 | 11 | 1872 | XP_016885129.1 | |
| XM_017029641.1 | 10 | 1794 | XP_016885130.1 | |
| XM_017029642.1 | 10 | 1675 | XP_016885131.1 | |
| XM_017029643.1 | 9 | 1650 | XP_016885132.1 | |
| XM_017029644.1 | 9 | 1599 | XP_016885133.1 | |
| XM_017029645.1 | 8 | 1572 | XP_016885134.1 | |
| XM_017029646.1 | 11 | 1499 | XP_016885135.1 | |
| AF230097.1 | 11 | 1590 | AAF73076.1 | |
| AF245664.1 | 11 | 1544 | AAF73428.1 | |
| AJ277724.1 | 11 | 1577 | CAB90213.1 | |
| BC050433.1 | 11 | 1568 | AAH50433.1 |
More Information
Set Membership: |
|

Genomic Map