Assay Details
Target Gene Details
Entrez Gene ID: | 2869 |
Gene Name: | G protein-coupled receptor kinase 5 |
Gene Aliases: |
GPRK5 |
Location: |
Chr.10:119207685-119455619 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRK5 | NM_005308.2 | NP_005299.1 | ||
| XM_005269707.1 | XP_005269764.1 | |||
| XM_005269708.1 | XP_005269765.1 | |||
| AK309761.1 | ||||
| AK313012.1 | ||||
| BC064506.1 | AAH64506.1 | |||
| L15388.1 | AAA58620.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv552214 | Chr.10:119100409 - 119209705 on Build GRCh38 | Loss |
|
| nsv1143523 | Chr.10:119206688 - 119207988 on Build GRCh38 | Deletion |
|
| nsv825584 | Chr.10:119167789 - 119229264 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map