Assay Details
Target Gene Details
Entrez Gene ID: | 54621 |
Gene Name: | V-set and immunoglobulin domain containing 10 |
Gene Aliases: |
- |
Location: |
Chr.12:118063593-118104402 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSIG10 | NM_019086.5 | 9 | 2740 | NP_061959.2 |
| XM_005253908.4 | 8 | 2093 | XP_005253965.1 | |
| XM_006719467.2 | 8 | 2437 | XP_006719530.1 | |
| XM_006719468.2 | 7 | 1832 | XP_006719531.1 | |
| XM_011538502.1 | 9 | 2737 | XP_011536804.1 | |
| XM_017019504.1 | 10 | 2754 | XP_016874993.1 | |
| XM_017019505.1 | 9 | 2360 | XP_016874994.1 | |
| XM_017019506.1 | 9 | 2874 | XP_016874995.1 | |
| AK000681.1 | BAA91322.1 | |||
| BC028698.2 | 9 | 2684 | AAH28698.2 | |
| BC034471.2 | 9 | 2679 | AAH34471.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581916 | Chr.12:117356518 - 118228514 on Build GRCh38 | Gain |
|
| nsv560389 | Chr.12:118064662 - 118126282 on Build GRCh38 | Gain |
|

Genomic Map