Assay Details
Target Gene Details
Entrez Gene ID: | 6898 |
Gene Name: | tyrosine aminotransferase |
Gene Aliases: |
- |
Location: |
Chr.16:71566851-71577095 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAT | NM_000353.2 | 5 | 577 | NP_000344.1 |
| AK313380.1 | 5 | 578 | ||
| BC130534.1 | 5 | 548 | ||
| X52520.1 | 5 | 574 | CAA36750.1 | |
| X55675.1 | 5 | 528 | CAA39210.1 |
More Information
Set Membership: |
|

Genomic Map