Assay Details
Target Gene Details
Entrez Gene ID: | 10170 |
Gene Name: | dehydrogenase/reductase 9 |
Gene Aliases: |
3-alpha-HSD, 3ALPHA-HSD, RDH-E2, RDH-TBE, RDH15, RDHL, RDHTBE, RETSDR8, SDR9C4 |
Location: |
Chr.2:169067035-169096167 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHRS9 | NM_001142270.1 | NP_001135742.1 | ||
| NM_001142271.1 | NP_001135743.1 | |||
| NM_001289763.1 | NP_001276692.1 | |||
| NM_199204.1 | NP_954674.1 | |||
| XM_011510475.1 | XP_011508777.1 | |||
| AF240697.1 | AAF82747.1 | |||
| AF240698.1 | AAF82748.1 | |||
| AF343729.1 | AAK37528.1 | |||
| AF529286.1 | AAN04008.1 | |||
| AF529287.1 | AAN04009.1 | |||
| AF529288.1 | AAN04010.1 | |||
| AK296625.1 | ||||
| AK313220.1 | ||||
| AY017349.1 | AAG49422.1 | |||
| AY359046.1 | AAQ89405.1 | |||
| BC058883.1 | AAH58883.1 | |||
| DB010228.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528457 | Chr.2:169036909 - 169297164 on Build GRCh38 | Gain |
|
| esv2762961 | Chr.2:169037369 - 169297835 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map