Assay Details
Target Gene Details
Entrez Gene ID: | 58472 |
Gene Name: | sulfide quinone reductase-like (yeast) |
Gene Aliases: |
CGI-44, PRO1975, SQOR |
Location: |
Chr.15:45631148-45691294 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SQRDL | NM_001271213.1 | 4 | 510 | NP_001258142.1 |
| NM_021199.3 | 3 | 703 | NP_067022.1 | |
| AF042284.1 | 3 | 393 | AAD41160.1 | |
| AF118085.1 | 2 | 224 | AAF22029.1 | |
| AF151802.1 | 3 | 387 | AAD34039.1 | |
| AK130140.1 | 4 | 510 | ||
| AK222463.1 | 3 | 402 | BAD96183.1 | |
| AK313111.1 | 3 | 382 | ||
| BC016836.1 | 3 | 358 | AAH16836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892684 | Chr.15:45307172 - 46327637 on Build GRCh38 | Gain |
|
| nsv1160292 | Chr.15:45351161 - 46175480 on Build GRCh38 | Duplication |
|
| esv3569553 | Chr.15:45366990 - 46080626 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map