Assay Details
Target Gene Details
Entrez Gene ID: | 11056 |
Gene Name: | DEAD-box helicase 52 |
Gene Aliases: |
HUSSY19, ROK1 |
Location: |
Chr.17:37609739-37643464 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX52 | NM_001291476.1 | 16 | 2098 | NP_001278405.1 |
| NM_007010.4 | 15 | 1983 | NP_008941.3 | |
| XM_011524232.2 | 15 | 1978 | XP_011522534.1 | |
| XM_011524233.2 | 13 | 1839 | XP_011522535.1 | |
| AB208938.1 | 13 | 3148 | BAD92175.1 | |
| AF077033.1 | 15 | 1950 | AAD27766.1 | |
| AJ010840.1 | 12 | 1402 | CAA09374.1 | |
| AK001245.1 | 12 | 1492 | ||
| AK001652.1 | 16 | 2079 | BAA91812.1 | |
| AK025721.1 | 5 | 485 | ||
| AK027425.1 | 12 | 1443 | ||
| BC041785.1 | 15 | 1965 | AAH41785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833431 | Chr.17:37530641 - 37726517 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map