Assay Details
Target Gene Details
Entrez Gene ID: | 6887 |
Gene Name: | TAL bHLH transcription factor 2 |
Gene Aliases: |
- |
Location: |
Chr.9:105662457-105663112 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAL2 | NM_005421.2 | 1 | 375 | NP_005412.1 |
| AA767775.1 | 1 | 282 | ||
| BC069422.1 | AAH69422.1 | |||
| BC126373.1 | ||||
| BC126375.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519023 | Chr.9:105609268 - 105692464 on Build GRCh38 | Gain |
|
| esv3891705 | Chr.9:104391828 - 106660921 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map