Assay Details
Target Gene Details
Entrez Gene ID: | 253982 |
Gene Name: | aspartate beta-hydroxylase domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.16:29900392-29919860 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASPHD1 | NM_181718.3 | 1 | 423 | NP_859069.2 |
| XM_017023107.1 | XP_016878596.1 | |||
| BC009033.1 | 1 | 150 | AAH09033.1 | |
| BC085011.1 | 1 | 271 | AAH85011.1 | |
| BC126319.1 | 1 | 391 | ||
| BC126321.1 | 1 | 391 | ||
| BC144152.1 | 1 | 391 | ||
| BU193236.1 | 1 | 423 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33251 | Chr.16:29900665 - 29901538 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map