Assay Details
Target Gene Details
Entrez Gene ID: | 112611 |
Gene Name: | RWD domain containing 2A |
Gene Aliases: |
RWDD2, dJ747H23.2 |
Location: |
Chr.6:83193313-83198935 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RWDD2A | NM_001322335.1 | NP_001309264.1 | ||
| NM_001322336.1 | 2 | 215 | NP_001309265.1 | |
| NM_001322337.1 | 2 | 215 | NP_001309266.1 | |
| NM_001322339.1 | NP_001309268.1 | |||
| NM_033411.4 | 2 | 215 | NP_219479.2 | |
| XM_017010222.1 | 2 | 179 | XP_016865711.1 | |
| XM_017010223.1 | XP_016865712.1 | |||
| AK295724.1 | ||||
| AK302978.1 | ||||
| AK309249.1 | 2 | 164 | ||
| AK313083.1 | 2 | 141 | ||
| BC010930.2 | 1 | 84 | AAH10930.1 | |
| BC104816.1 | AAI04817.1 | |||
| BC112003.1 | AAI12004.1 | |||
| BG676907.1 | 2 | 164 | ||
| CD108545.1 | 2 | 136 | ||
| DC377982.1 | 2 | 215 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026095 | Chr.6:83040907 - 83209453 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map