Assay Details
Target Gene Details
Entrez Gene ID: | 5511 |
Gene Name: | protein phosphatase 1 regulatory subunit 8 |
Gene Aliases: |
ARD-1, ARD1, NIPP-1, NIPP1, PRO2047 |
Location: |
Chr.1:27830741-27851672 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R8 | NM_002713.3 | 6 | 1454 | NP_002704.1 |
| NM_014110.4 | 7 | 1273 | NP_054829.2 | |
| NM_138558.2 | 7 | 1555 | NP_612568.1 | |
| XM_017001594.1 | 6 | 974 | XP_016857083.1 | |
| AF061958.1 | 7 | 1201 | AAD31541.1 | |
| AF061959.1 | 7 | 1356 | AAD31542.1 | |
| AF126488.1 | 7 | 1189 | AAD22486.1 | |
| AK297771.1 | ||||
| AX775985.1 | 7 | 1189 | CAE11692.1 | |
| BC001597.1 | 6 | 1081 | AAH01597.1 | |
| BC013360.2 | 6 | 1131 | AAH13360.1 | |
| BC093017.1 | 7 | 1193 | AAH93017.1 | |
| DQ223931.1 | ABB04281.1 | |||
| U14575.1 | 6 | 1441 | AAA64749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509069 | Chr.1:27817460 - 27905956 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map