Assay Details
Target Gene Details
Entrez Gene ID: | 8266 |
Gene Name: | ubiquitin like 4A |
Gene Aliases: |
DX254E, DXS254E, G6PD, GDX, GET5, MDY2, TMA24, UBL4 |
Location: |
Chr.X:154483717-154486670 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBL4A | NM_014235.4 | 4 | 1666 | NP_055050.1 |
| AK000405.1 | 4 | 1601 | ||
| BC043346.2 | 4 | 1593 | AAH43346.1 | |
| BC053589.1 | 4 | 1627 | AAH53589.1 | |
| CK826435.1 | 1 | 499 | ||
| M35604.1 | 1 | 159 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
| nsv519042 | Chr.X:154082978 - 154601444 on Build GRCh38 | Gain |
|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map