Assay Details
Target Gene Details
Entrez Gene ID: | 1007 |
Gene Name: | cadherin 9 |
Gene Aliases: |
- |
Location: |
Chr.5:26880600-27038582 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH9 | NM_016279.3 | 11 | 1915 | NP_057363.3 |
| AB035302.1 | 11 | 1900 | BAA87416.1 | |
| AK294183.1 | 4 | 762 | ||
| AK307779.1 | 11 | 1916 | ||
| BC107594.1 | 11 | 1877 | AAI07595.1 | |
| BC113745.1 | 11 | 1877 | AAI13746.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759335 | Chr.5:26755188 - 26934733 on Build GRCh38 | Loss |
|
| esv3604457 | Chr.5:26834588 - 26890217 on Build GRCh38 | Gain |
|
| nsv597645 | Chr.5:26540207 - 28477865 on Build GRCh38 | Gain |
|
| nsv597679 | Chr.5:26872627 - 26919239 on Build GRCh38 | Loss |
|
| nsv462024 | Chr.5:26874532 - 26900794 on Build GRCh38 | Loss |
|
| nsv1022430 | Chr.5:26664643 - 26945247 on Build GRCh38 | Gain |
|
| dgv9699n54 | Chr.5:26865192 - 26977247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map