Assay Details
Target Gene Details
Entrez Gene ID: | 63875 |
Gene Name: | mitochondrial ribosomal protein L17 |
Gene Aliases: |
L17mt, LIP2, MRP-L17, MRP-L26, RPL17L, RPML26 |
Location: |
Chr.11:6680385-6683401 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL17 | NM_022061.3 | 3 | 693 | NP_071344.1 |
| AF141338.1 | 3 | 615 | AAG49441.1 | |
| AF164797.1 | 3 | 608 | AAF80761.1 | |
| AK026857.1 | 3 | 598 | BAB15575.1 | |
| BC012306.1 | 3 | 594 | AAH12306.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035138 | Chr.11:6607812 - 6980555 on Build GRCh38 | Gain |
|
| nsv832060 | Chr.11:6663417 - 6853326 on Build GRCh38 | Gain |
|
| nsv7658 | Chr.11:6660591 - 6693515 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map