Assay Details
Target Gene Details
Entrez Gene ID: | 57017 |
Gene Name: | coenzyme Q9 |
Gene Aliases: |
C16orf49, COQ10D5 |
Location: |
Chr.16:57447425-57461275 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COQ9 | NM_020312.3 | NP_064708.1 | ||
| AF161444.1 | AAF29004.1 | |||
| AK290627.1 | ||||
| AL136884.1 | CAB66818.2 | |||
| BC001478.2 | AAH01478.2 | |||
| BC054340.1 | AAH54340.2 | |||
| BC064946.1 | AAH64946.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833245 | Chr.16:57434073 - 57584619 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map