Assay Details
Target Gene Details
Entrez Gene ID: | 5655 |
Gene Name: | kallikrein related peptidase 10 |
Gene Aliases: |
NES1, PRSSL1 |
Location: |
Chr.19:51012744-51020175 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK10 | NM_001077500.1 | NP_001070968.1 | ||
| NM_002776.4 | NP_002767.2 | |||
| XM_006723289.3 | XP_006723352.1 | |||
| AK292365.1 | ||||
| BC002710.2 | AAH02710.1 | |||
| BM830577.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2677627 | Chr.19:51015389 - 51020436 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map