Assay Details
Target Gene Details
Entrez Gene ID: | 23645 |
Gene Name: | protein phosphatase 1 regulatory subunit 15A |
Gene Aliases: |
GADD34 |
Location: |
Chr.19:48872392-48876062 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R15A | NM_014330.3 | NP_055145.3 | ||
| AK001361.1 | BAA91649.1 | |||
| AK225632.1 | ||||
| AK296668.1 | ||||
| AK297615.1 | ||||
| AK298765.1 | ||||
| BC003067.2 | AAH03067.1 | |||
| CR457259.1 | CAG33540.1 | |||
| U83981.1 | AAC25631.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv470148 | Chr.19:48865586 - 48906934 on Build GRCh38 | Gain |
|
| nsv833856 | Chr.19:48847866 - 48986483 on Build GRCh38 | Loss |
|
| nsv833855 | Chr.19:48750098 - 48949781 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77919834] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map