Assay Details
Target Gene Details
Entrez Gene ID: | 91181 |
Gene Name: | nucleoporin 210 like |
Gene Aliases: |
- |
Location: |
Chr.1:153992582-154155116 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 32 - Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUP210L | NM_001159484.1 | NP_001152956.1 | ||
| NM_207308.2 | NP_997191.2 | |||
| XM_011510122.1 | XP_011508424.1 | |||
| XM_011510123.1 | XP_011508425.1 | |||
| XM_011510124.1 | XP_011508426.1 | |||
| XM_017002788.1 | XP_016858277.1 | |||
| XM_017002789.1 | XP_016858278.1 | |||
| AK125924.1 | BAC86345.1 | |||
| AK128108.1 | BAC87279.1 | |||
| AL133633.1 | CAB63757.1 | |||
| AL833964.1 | CAD38812.1 | |||
| BX648132.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3578197 | Chr.1:154013254 - 154028290 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map