Assay Details
Target Gene Details
Entrez Gene ID: | 81628 |
Gene Name: | TSC22 domain family member 4 |
Gene Aliases: |
THG-1, THG1, TILZ2 |
Location: |
Chr.7:100466519-100479279 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D4 | NM_001303043.1 | NP_001289972.1 | ||
| NM_030935.4 | NP_112197.1 | |||
| NR_130118.1 | ||||
| NR_130119.1 | ||||
| AJ133115.1 | CAB43491.1 | |||
| AK296580.1 | ||||
| AK297702.1 | ||||
| AK314824.1 | ||||
| BC001486.2 | AAH01486.1 | |||
| BC001966.1 | AAH01966.1 | |||
| BC002972.1 | AAH02972.1 | |||
| BC010406.2 | AAH10406.1 | |||
| BC031622.1 | 3 | 1561 | AAH31622.1 | |
| BX341171.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
| nsv527050 | Chr.7:100458543 - 100528352 on Build GRCh38 | Loss |
|
| nsv831071 | Chr.7:100320402 - 100515790 on Build GRCh38 | Loss |
|
| nsv951385 | Chr.7:100467278 - 100496477 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map