Assay Details
Target Gene Details
Entrez Gene ID: | 54829 |
Gene Name: | asporin |
Gene Aliases: |
OS3, PLAP-1, PLAP1, SLRR1C |
Location: |
Chr.9:92456205-92482562 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASPN | NM_001193335.1 | 6 | 1118 | NP_001180264.1 |
| NM_017680.4 | 8 | 1351 | NP_060150.4 | |
| AF316824.1 | 7 | 1069 | AAK35161.1 | |
| AK000136.1 | 6 | 721 | BAA90967.1 | |
| AK027359.1 | 6 | 921 | BAB55060.1 | |
| AK172737.1 | 6 | 821 | ||
| AK304501.1 | 6 | 925 | ||
| AK308107.1 | 8 | 1265 | ||
| AY029191.1 | 8 | 1284 | AAK31800.1 | |
| AY358329.1 | 8 | 1284 | AAQ88695.1 | |
| BC063114.1 | 8 | 1316 | AAH63114.1 |
Target Gene Details
Entrez Gene ID: | 401541 |
Gene Name: | centromere protein P |
Gene Aliases: |
CENP-P |
Location: |
Chr.9:92325457-92615164 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPP | NM_001012267.2 | NP_001012267.1 | ||
| NM_001286969.1 | NP_001273898.1 | |||
| XM_011518689.1 | XP_011516991.1 | |||
| AK092603.1 | ||||
| AK307435.1 | ||||
| BC071726.1 | AAH71726.1 | |||
| BX537851.1 | CAD97859.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049599 | Chr.9:92407857 - 92503276 on Build GRCh38 | Loss |
|
| esv3891693 | Chr.9:92163398 - 92538277 on Build GRCh38 | Gain |
|
| nsv1047337 | Chr.9:92343356 - 92585202 on Build GRCh38 | Loss |
|
| esv3621080 | Chr.9:92435546 - 92479530 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map