Assay Details
Target Gene Details
Entrez Gene ID: | 3551 |
Gene Name: | inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta |
Gene Aliases: |
IKK-beta, IKK2, IKKB, IMD15, NFKBIKB |
Location: |
Chr.8:42271302-42332653 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IKBKB | NM_001190720.2 | 1 | 56 | NP_001177649.1 |
| NM_001242778.1 | 1 | 56 | NP_001229707.1 | |
| NM_001556.2 | 1 | 56 | NP_001547.1 | |
| NR_033818.1 | 1 | 56 | ||
| NR_033819.1 | 1 | 56 | ||
| NR_040009.1 | 1 | 56 | ||
| XM_005273490.2 | 1 | 56 | XP_005273547.1 | |
| XM_005273491.4 | 1 | 51 | XP_005273548.1 | |
| XM_005273492.3 | 1 | 56 | XP_005273549.1 | |
| XM_005273493.3 | XP_005273550.1 | |||
| XM_005273494.2 | 1 | 56 | XP_005273551.1 | |
| XM_005273496.3 | XP_005273553.1 | |||
| XM_005273498.3 | XP_005273555.1 | |||
| XM_011544518.2 | 1 | 56 | XP_011542820.1 | |
| XM_011544521.2 | XP_011542823.1 | |||
| XM_011544522.2 | XP_011542824.1 | |||
| XM_017013396.1 | XP_016868885.1 | |||
| AB209090.1 | BAD92327.1 | |||
| AF080158.1 | AAD08997.1 | |||
| AK091297.1 | ||||
| AK298241.1 | ||||
| AK302723.1 | ||||
| AK303294.1 | ||||
| AK303528.1 | ||||
| AK316418.1 | ||||
| AL708460.1 | 1 | 55 | ||
| BC006231.2 | AAH06231.1 | |||
| BC108694.1 | AAI08695.1 | |||
| DA043516.1 | 1 | 56 | ||
| DA105584.1 | 1 | 56 | ||
| DA159382.1 | 1 | 56 | ||
| DA709000.1 | 1 | 56 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831301 | Chr.8:42244080 - 42473666 on Build GRCh38 | Loss |
|
| nsv508508 | Chr.8:42256602 - 42355479 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map