Assay Details
Target Gene Details
Entrez Gene ID: | 8669 |
Gene Name: | eukaryotic translation initiation factor 3 subunit J |
Gene Aliases: |
EIF3S1, eIF3-alpha, eIF3-p35 |
Location: |
Chr.15:44537068-44562803 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3J | NM_001284335.1 | 7 | 1816 | NP_001271264.1 |
| NM_001284336.1 | 6 | 1831 | NP_001271265.1 | |
| NM_003758.3 | 8 | 1978 | NP_003749.2 | |
| AK002006.1 | 8 | 1898 | ||
| BU621047.1 | 1 | 597 | ||
| U97670.1 | 9 | 1845 | AAC78729.1 |
More Information
Set Membership: |
|

Genomic Map