Assay Details
Target Gene Details
Entrez Gene ID: | 5701 |
Gene Name: | proteasome 26S subunit, ATPase 2 |
Gene Aliases: |
MSS1, Nbla10058, S7 |
Location: |
Chr.7:103344405-103369395 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMC2 | NM_001204453.1 | NP_001191382.1 | ||
| NM_002803.3 | NP_002794.1 | |||
| XM_005250505.1 | XP_005250562.1 | |||
| AB075520.1 | BAE45763.1 | |||
| AK298529.1 | ||||
| AK298821.1 | ||||
| AK312648.1 | ||||
| BC002589.2 | AAH02589.1 | |||
| BE269755.1 | ||||
| BM754312.1 | ||||
| D11094.1 | BAA01868.1 |
Target Gene Details
Entrez Gene ID: | 375611 |
Gene Name: | solute carrier family 26 member 5 |
Gene Aliases: |
DFNB61, PRES |
Location: |
Chr.7:103352598-103446177 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC26A5 | NM_001321787.1 | NP_001308716.1 | ||
| NM_206883.2 | NP_996766.1 | |||
| NM_206884.2 | NP_996767.1 | |||
| NM_206885.2 | NP_996768.1 | |||
| NR_135802.1 | ||||
| AY256823.1 | AAP31532.1 | |||
| AY256824.1 | AAP31533.1 | |||
| AY256825.1 | AAP31534.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5884 | Chr.7:103321719 - 103357594 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map