Assay Details
Target Gene Details
Entrez Gene ID: | 80004 |
Gene Name: | epithelial splicing regulatory protein 2 |
Gene Aliases: |
RBM35B |
Location: |
Chr.16:68228547-68236233 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESRP2 | NM_024939.2 | 15 | 2429 | NP_079215.2 |
| XM_005256153.4 | 15 | 2368 | XP_005256210.1 | |
| XM_006721273.3 | 15 | 2437 | XP_006721336.1 | |
| XM_011523338.2 | 14 | 2329 | XP_011521640.1 | |
| AF289577.1 | 9 | 1531 | AAL55761.1 | |
| AK025571.1 | 15 | 2358 | BAB15173.1 | |
| AK025901.1 | 4 | 694 | BAB15275.1 | |
| BC030146.2 | 15 | 2251 | AAH30146.1 |
More Information
Set Membership: |
|

Genomic Map